Learn More
Research Products International Corp Nalgene Rectangular Poly Bottle w/Closure, 2gal
SDP
Catalog No. 501977896
Encompass
Easily fill, transfer and clean HDPE Rectangular Carboys without Spigots using the wide-mouth opening. Featuring a sturdy, space-saving design, these high density polyethylene carboys have convenient molded-in graduations to deliver in liters and gallons.
Catalog No. 50-197-7896 Supplier Research Products International Corp Supplier No. 22110020
Easily fill, transfer and clean HDPE Rectangular Carboys without Spigots using the wide-mouth opening. Featuring a sturdy, space-saving design, these high density polyethylene carboys have convenient molded-in graduations to deliver in liters and gallons.