Learn More
Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge
GREENER_CHOICE
Catalog No. 01071 Shop All Thermo Scientific Products
Change view
Click to view available options
Flow Rate:
15 cc/stroke
36 cc/stroke
For Use With:
6132-2000 Repair Kit
6132-3000 Repair Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate For Use With
01-071 15 cc/stroke 6132-2000 Repair Kit
01-071A 36 cc/stroke 6132-3000 Repair Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-071 Supplier Thermo Scientific™ Supplier No. 61310010
Add to Cart
Add to Cart
Quickly attain and hold vacuum of 25 in. (635 mm) Hg with these hand-operated vacuum pumps featuring a new, more durable and ergonomic handle design.
Quickly attain and hold vacuum of 25 in. (635 mm) Hg with Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge. These pumps feature a new, more durable and ergonomic handle design. This repairable economy version is ideal for field or lab use.
Easy and Convenient Operation
  • Polyvinyl Chloride (PVC) body
  • Ergonomic vacuum trigger release requires only one hand to operate and releases vacuum with the touch of an index finger
  • Adjustable vacuum release rate provides control
  • Full blank port pressure is obtained with only two strokes, making operation efficient
  • Removable cover on exhaust port and trigger to release vacuum without disconnecting pumps from the line
  • Economical design without vacuum gauge 
  • Adjustable vacuum release rate controlled by trigger on pump
  • Nozzle fits standard 0.25″ I.D. tubing
  • Can be disassembled for repair and replacement of internal parts (see catalog numbers for repair kits in the table below)
Includes:
Supplied with 2 ft. length of Clear Plastic Tubing
TRUSTED_SUSTAINABILITY
Specifications
Description Repairable Hand Vacuum Pump
No. per Pack 1
No. per Case 4
Max. Vacuum 25 in. Hg
Flow Rate 15 cc/stroke
Type Vacuum Pump
For Use With 6132-2000 Repair Kit