Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Septum Closures: Sterile, Bulk Pack, 38-430mm
GREENER_CHOICE

Catalog No. 0292330 Shop All Thermo Scientific Products
Change view
Click to view available options
Neck Finish:
38-430
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
02-923-30 38-430
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-923-30 Supplier Thermo Scientific™ Supplier No. 3421780384
  • $1,273.00
    $780.65 / Case of 240

    Save $492.35

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Combine elastomeric sealing of silicone with nonreactive PTFE reagent contact surfaces in these septum closures for media bottles.

Combine the elastomeric sealing of silicone with nonreactive polytetrafluoroethylene reagent contact surfaces in Thermo Scientific™ Nalgene™ Septum Closures, for use with Nalgene™ Sterile PET & PETG media bottles.

Features:
  • Sterile 10-6 SAL
  • No adhesives used to attach septum to high density polyethylene closure
  • Non-cytotoxic and non-pyrogenic
  • Dual septum material for improved gas barrier

Ordering Information:

  • Closures are bulk packed for easy handling.
  • Order Nalgene PET or PETG bottles without closures separately.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Nalgene Closure for Media Bottles
Material HDPE
No. per Case 240
No. per Pack 240
Packaging Bulk
Sterility Sterile
Liner Material Silicone, PTFE
Neck Finish 38-430
Fit for Purpose Sterile
For Use With (Equipment) Nalgene Sterile PET and PETG media bottles
Closure Size 38-430 mm
Certifications/Compliance Manufactured in compliance with ISO 13485 quality requirements
Product Type Septum Closure
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.