Learn More
Thermo Scientific™ Nalgene™ PETG Ultra-Low Particulate Media Bottles
Shop All Thermo Scientific Products

Description
Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Eliminates costly washing, depyrogenation and autoclaving steps
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
- Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
- Supported by extensive validation documentation
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Clear |
| Diameter (English) | 2.9 in. |
| Diameter (English) Inner Neck | 1.10 in. |
| Diameter (Metric) Inner Neck | 28 mm |
| For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
| Height (Metric) With Closure | 177 mm |
| Material | PETG |
| No. per Case | 70/Cs. |
| No. per Pack | 5/Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.