Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays

Catalog No. 0992315BLC Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-923-15BLC 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-923-15BLC Supplier Thermo Scientific™ Supplier No. 3220200500LC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Package and ship liquid medias, buffers and sera using Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles offer excellent gas barrier properties.

Package and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.

  • Heavy-walled and durable
  • Shatter-resistant
  • Provide excellent gas barrier properties
  • In shrink-wrap trays
  • Molded in graduations
  • Clear bottles for easy identification of bottles
  • Non-sterile
  • Leakproof

Note:

The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Clear
Description 500 mL, 38-430 closure, tray Pk.
Diameter (English) 2.9 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Material PETG
No. per Case 40
No. per Pack 20
Sterility Non-sterile
Mouth Narrow
Height (Metric) 177 mm
Height (English) 6.97 in.
Closure Material HDPE
Closure Size 38-430 mm
Diameter (Metric) 74 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.