Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays



Description
Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.
- Heavy-walled and durable
- Shatter-resistant
- Provide excellent gas barrier properties
- In shrink-wrap trays
- Molded in graduations
- Clear bottles for easy identification of contents
- Nonsterile
- Leakproof†
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8.5 oz. |
| Color | Clear |
| Description | 250mL, 38-430 closure, tray pack |
| Diameter (English) | 2.4 in. |
| Diameter (English) Inner Neck | 1.1 in. |
| Diameter (Metric) Inner Neck | 28 mm |
| For Use With (Application) | Storing Media and Sera |
| Material | PETG |
| No. per Case | 60 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.