Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 14178168 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
53B
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
14-178-168 60 mL 24-415 mm
02-923-82 30 mL 20-415 mm
02-923-155 125 mL 38-430 mm
03-313-34D 250 mL 38-430 mm
09-923-15B 500 mL 38-430 mm
03-313-34F 1 L 38-430 mm
03-313-34G 2 L 53B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-178-168 Supplier Thermo Scientific™ Supplier No. 3220200060
  • $682.00
    $468.65 / Case of 200

    Save $213.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.

Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.

  • Heavy-walled and durable
  • Shatter-resistant
  • Provide excellent gas barrier properties
  • In shrink-wrap trays
  • Molded in graduations
  • Clear bottles for easy identification of contents
  • Nonsterile
  • Leakproof†

Note:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 60 mL
Color Clear
Description 60mL, 24-415 closure, tray pack
Diameter (English) 1.6 in.
Diameter (English) Inner Neck 0.71 in.
Diameter (Metric) Inner Neck 18 mm
For Use With (Application) Storing Media and Sera
Material PETG
No. per Case 200
No. per Pack 40
Sterility Non-sterile
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 82 mm
Height (English) 3.2 in.
Closure Material HDPE
Closure Size 24-415 mm
Diameter (Metric) 41 mm
Capacity (English) 2 oz.
Shape Square
Certifications/Compliance USP Class VI, FDA Compliance 21 CFR
Product Type Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.