Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
GSA_VAGREENER_CHOICE

Catalog No. 03007 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-007 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-007 Supplier Thermo Scientific™ Supplier No. 24140030
  • $736.50
    $549.65 / Case of 4

    Save $186.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dispense strong acids/alkalis, reagents and solvents with these FEP drop-dispensing bottles which feature excellent chemical resistance for dispensing virtually any chemical.

Dispense strong acids/alkalis, reagents and solvents with Thermo Scientific™ Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™, featuring excellent chemical resistance for dispensing virtually any chemical. Tethered spout cap snaps securely into place and won't pop off or leak when the closed bottle is squeezed. Usable over a wide temperature range -105°C to +150°C. Leakproof† when used with provided Nalgene closure and autoclavable.

Features:
  • Dispenses only one drop at a time for dependable delivery
  • Convenient one-handed operation frees up your other hand
  • Transparent so you can easily view liquid levels
  • Includes ETFE dropping closure and cap

Ordering Information:

One per package; case of four.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 mL
Color White
Description Teflon FEP Drop-Dispensing Bottle with Dropping Closure and Cap
Material FEP
No. per Case 4
No. per Pack 1
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 99.06 mm
Height (English) 3.9 in.
Closure Material ETFE
Diameter (Metric) 34.29 mm
Closure Type Dropping Closure and Cap
Through the Closure Dispensing Yes
Autoclavable Autoclavable
Capacity (English) 1 oz.
Shape Round
Certifications/Compliance USP Class VI
Product Type Dropping Bottle
Show More Show Less
Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?

Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.