Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene Torque Fitting Tool for Nalgene Bottle and Closure Combinations

Catalog No. 11100285 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
13 mm Closures
20 mm Closures
20 mm Serum Vial Closures, HDPE
24 mm Closures
28 mm Closures
38 mm Biotainer Closures
38-415 Closures
38-430 Closures
43 mm Closures
48 mm Biotainer Closures
48 mm Closures
53 mm Closures
53B Closures
63 mm Closures
83B Closures
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-100-285 38-415 Closures
11-100-281 13 mm Closures
11-100-282 20 mm Closures
11-100-283 24 mm Closures
11-100-284 28 mm Closures
11-100-286 43 mm Closures
11-100-272 48 mm Closures
11-100-273 53 mm Closures
11-100-274 63 mm Closures
11-100-275 53B Closures
11-100-276 83B Closures
11-100-277 38-430 Closures
11-100-278 20 mm Serum Vial Closures, HDPE
11-100-279 38 mm Biotainer Closures
11-100-280 48 mm Biotainer Closures
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 11-100-285 Supplier Thermo Scientific™ Supplier No. 31950038
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensure a guaranteed leak-proof seal with proper torque settings.

The Nalgene Bottle Torque Fittings represent a specialized closure solution engineered to deliver precise torque application and guaranteed leakproof performance across Nalgene's laboratory vessel portfolio. These fittings are uniquely designed for compatibility with various Nalgene bottle and closure configurations, including lab bottles, packaging bottles, media bottles, biotainers, and PETG serum vials.

  • Comprehensive Size Range: Available in multiple sizes engineered to accommodate the majority of standard laboratory bottles and closure configurations
  • Multi-Material Closure Compatibility: Designed for optimal performance with both high-density polyethylene (HDPE) and polypropylene (PP) closure materials
  • Nalgene-Specific Design: Custom-engineered geometry specifically optimized for Nalgene laboratory closure systems
  • Universal Torque Tool Integration: Compatible with industry-standard 1/4 in. and 3/8 in. square drive torque tools for precise torque application
  • Manual and Automated Operation: Supports both manual torque application and automated torque systems for laboratory workflow integration
  • Product Specifications and Operating Parameters - Detailed torque settings are available in the documents section of this page.
  • Comprehensive Documentation Provides Essential Operating Procedures - Detailed operational instructions are provided in the accompanying documentation section to ensure proper implementation and use of the bottle and closure system.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Torque Fitting
For Use With (Application) Proper torque of closures
For Use With (Equipment) 38-415 Closures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.