Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures
Catalog No. 14178150
Change view
Click to view available options
Closure Size:
20 to 415 mm
38 to 430 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
14-178-150 20 to 415 mm
14-178-158 38 to 430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-178-150 Supplier Thermo Scientific™ Supplier No. 21952020
Assure accurate torque readings when applying closures onto bottles with Nalgene™ Torque Wrench Fittings for ETFE closures.
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures. Proper application torque is essential for leakproof† performance.
  • Durable epoxy resin construction
  • Specifically designed for Nalgene ETFE closures
Compatible with:
Compatible with Thermo Scientific Nalgene torque wrench 2195-6169 † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Torque Wrench Fittings
Closure Size 20 to 415 mm
Drive Size 1/4 in.
For Use With (Equipment) Nalgene Torque Wrenches (Cat. No. 2195-6169)
Material Epoxy
No. per Pack 1 Ea.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.