Learn More
Research Products International Corp Nalidixic Acid 1 KG
SDP
Catalog No. 502230536
Encompass_Preferred
Nalidixic Acid Sodium Salt is a chemical compound that is used for its antimicrobial properties and has various applications in microbiology, and pharmaceutical research. Nalidixic Acid Sodium Salt is employed as a reference standard and selective agent in microbiological culture media.
Researchers use it to assess the effectiveness of antibiotics against different bacterial strains.
In pharmaceutical research, Nalidixic Acid Sodium Salt can be used as a reference standard for antibiotic testing and formulation. It supports the development and optimization of antibiotics and strategies to combat antibiotic-resistant bacteria. 
WARNING! This product can expose you to Nalidixic acid, a chemical which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
I
Catalog No. 50-223-0536 Supplier Research Products International Corp Supplier No. N420001000.0
Add to Cart
Add to Cart
Nalidixic Acid Sodium Salt is a chemical compound that is used for its antimicrobial properties and has various applications in microbiology, and pharmaceutical research. Nalidixic Acid Sodium Salt is employed as a reference standard and selective agent in microbiological culture media.
Researchers use it to assess the effectiveness of antibiotics against different bacterial strains.
In pharmaceutical research, Nalidixic Acid Sodium Salt can be used as a reference standard for antibiotic testing and formulation. It supports the development and optimization of antibiotics and strategies to combat antibiotic-resistant bacteria. 
WARNING! This product can expose you to Nalidixic acid, a chemical which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
I