Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nalidixic acid | 389-08-2 | MFCD00006884 | 100.0% | 10 MG
SDP

Catalog No. 5000298928
Encompass_Preferred

Nalidixic acid analytical standard is a quinolone antibiotic reference material supplied for research, analytical testing, and quality control. It inhibits DNA gyrase and topoisomerase IV, blocking DNA replication in susceptible bacteria.

  • High purity (99.98%).
  • Chemical formula C12H12N2O3; molecular weight 232.24 g/mol.
  • CAS number 389-08-2 for unambiguous identification.
  • Supplied as a 10 MG analytical standard suitable for research and QC.
  • Intended for analytical and quality-control applications.
  • Store under conditions specified in the certificate of analysis.

Catalog No. 50-002-98928 Supplier Medchemexpress LLC Supplier No. HYB0398R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nalidixic acid analytical standard is a quinolone antibiotic reference material supplied for research, analytical testing, and quality control. It inhibits DNA gyrase and topoisomerase IV, blocking DNA replication in susceptible bacteria.

  • High purity (99.98%).
  • Chemical formula C12H12N2O3; molecular weight 232.24 g/mol.
  • CAS number 389-08-2 for unambiguous identification.
  • Supplied as a 10 MG analytical standard suitable for research and QC.
  • Intended for analytical and quality-control applications.
  • Store under conditions specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.