Learn More
Medchemexpress LLC Nalidixic acid | 389-08-2 | MFCD00006884 | 100.0% | 25 MG
SDP
Catalog No. 5000450616
Encompass_Preferred
Nalidixic acid (Standard) is the analytical standard of Nalidixic acid, intended for research and analytical applications. It is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria. Nalidixic acid exhibits bacteriostatic action at lower concentrations and is bactericidal at higher concentrations. It works by inhibiting a subunit of DNA gyrase and topoisomerase IV, thereby reversibly blocking DNA replication in susceptible bacteria.
  • Analytical standard for nalidixic acid.
  • Quinolone antibiotic.
  • Effective against gram-positive and gram-negative bacteria.
  • Bacteriostatic at lower concentrations, bactericidal at higher concentrations.
  • Inhibits a subunit of DNA gyrase and topoisomerase IV.
  • Reversibly blocks DNA replication in susceptible bacteria.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-004-50616 Supplier Medchemexpress LLC Supplier No. 25MGHYB0398R
Nalidixic acid (Standard) is the analytical standard of Nalidixic acid, intended for research and analytical applications. It is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria. Nalidixic acid exhibits bacteriostatic action at lower concentrations and is bactericidal at higher concentrations. It works by inhibiting a subunit of DNA gyrase and topoisomerase IV, thereby reversibly blocking DNA replication in susceptible bacteria.
  • Analytical standard for nalidixic acid.
  • Quinolone antibiotic.
  • Effective against gram-positive and gram-negative bacteria.
  • Bacteriostatic at lower concentrations, bactericidal at higher concentrations.
  • Inhibits a subunit of DNA gyrase and topoisomerase IV.
  • Reversibly blocks DNA replication in susceptible bacteria.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.