Learn More
Research Products International Corp Nalidixic Acid, 5 Grams
SDP
Supplier:  Research Products International Corp N420005.0
Encompass_Preferred
Nalidixic Acid Sodium Salt is a chemical compound that is used for its antimicrobial properties and has various applications in microbiology, and pharmaceutical research. Nalidixic Acid Sodium Salt is employed as a reference standard and selective agent in microbiological culture media. It inhibits the growth of susceptible bacteria, making it a valuable tool for studying bacterial susceptibility and resistance patterns. 
Researchers use it to assess the effectiveness of antibiotics against different bacterial strains, contributing to our understanding of microbial biology and antibiotic resistance. 
WARNING! This product can expose you to Nalidixic acid, a chemical which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
I
Catalog No. 50-488-853