Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalidixic acid, 99.5%

Catalog No. AC169900250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169900250 25 g Glass bottle
AC169900050 5 g Glass bottle
AC169901000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169900250 Supplier Thermo Scientific Chemicals Supplier No. 169900250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Nalixidic acid, or nalixidin, CAS # 389-08-2, is a quinolone antibiotic used to inhibit DNA gyrase and prevent cell division. | CAS: 389-08-2 | C12H12N2O3 | 232.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nalixidic acid is a synthetic quinolone that is a monocarboxylic acid composed of 1,8-naphthyridine substituted with carboxylic acid, ethyl, and methyl groups

• Nalixidic acid is a DNA gyrase inhibitor that prevents DNA synthesis and subsequent cell divisions

Applications

• Nalixidic acid is primarily effective against gram-negative bacteria

• It may be useful for bacterial resistance studies and laboratory research on drug allergies and cross-reactivity

• Nalixidic acid is suitable for high-performance liquid chromatography (HPLC) and related biochemistry laboratory applications

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389-08-2
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.24
MDL Number MFCD00006884
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
PubChem CID 4421
ChEBI CHEBI:100147
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 225.0°C to 231.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Glass bottle
Quantity 25 g
Beilstein 25, V,7, 384
Merck Index 15, 6444
Solubility Information Solubility in water: 0.1g/L (23°C). Other solubilities: soluble in alkali solutions,35mg/mL chloroform- 1.6mg/mL toluene (23°C),1.3mg/mL methanol- 0.9mg/mL ethanol (23°C),0.1mg/mL ether (23°C)
Formula Weight 232.24
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material Nalidixic acid, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
EINECSNumber 206-864-7
RTECSNumber QN2885000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.