Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalidixic Acid, G-Biosciences
SCI_ED

Catalog No. S139785
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139785 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139785 Supplier G-Biosciences Supplier No. RC1060
Add to Cart
Add to Cart

Nalidixic acid is a naphthyridone antibiotic that is similar in structure and mechanism to quinolones and is effective against both gram-positive and gram-negative bacteria.

It acts as a bacteriostatic in lower concentrations but can be bacteriocidal at higher concentrations. Nalidixic acid indirectly inhibits proteins synthesis by binding DNA and interfering with enzymes involved in DNA replication and transcription.

Chemical Identifiers

Molecular Weight (g/mol) 232.24 g/mol

Specifications

Boiling Point 413.1°C
Quantity 100 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.