Learn More
Research Products International Corp Nalidixic Acid Sodm Salt 500 G
SDP
Catalog No. 502230538
Encompass_Preferred
Nalidixic Acid Sodium Salt is a compound of notable importance in microbiology and pharmaceutical research. This sodium salt formulation of nalidixic acid is a valuable tool for investigating bacterial resistance mechanisms and antibiotic development. 
In microbiology, Nalidixic Acid Sodium Salt is widely employed to study bacterial susceptibility and resistance patterns. It serves as a selective agent in microbiological culture media, inhibiting the growth of susceptible bacteria. Researchers use it to assess the effectiveness of antibiotics and to understand the molecular mechanisms behind bacterial resistance, contributing to our understanding of microbial biology and antibiotic resistance. 
Pharmaceutical researchers also rely on Nalidixic Acid Sodium Salt to develop new antibiotics and optimize existing ones.
Catalog No. 50-223-0538 Supplier Research Products International Corp Supplier No. N23100500.0
Nalidixic Acid Sodium Salt is a compound of notable importance in microbiology and pharmaceutical research. This sodium salt formulation of nalidixic acid is a valuable tool for investigating bacterial resistance mechanisms and antibiotic development. 
In microbiology, Nalidixic Acid Sodium Salt is widely employed to study bacterial susceptibility and resistance patterns. It serves as a selective agent in microbiological culture media, inhibiting the growth of susceptible bacteria. Researchers use it to assess the effectiveness of antibiotics and to understand the molecular mechanisms behind bacterial resistance, contributing to our understanding of microbial biology and antibiotic resistance. 
Pharmaceutical researchers also rely on Nalidixic Acid Sodium Salt to develop new antibiotics and optimize existing ones.