Learn More
Selleck Chemical LLC Nalidixic acid sodium salt
SDP
Supplier:  Selleck Chemical LLC E1905250MG
Encompass_Preferred
Nalidixic acid sodium salt is an inhibitor of bacterial DNA gyrase and topoisomerase IV essential enzymes for DNA supercoiling during replication making it an effective quinolone antibiotic against both gram-positive and gram-negative bacteria
Catalog No. 50-313-7803