Learn More
Alkali Scientific Nalidixic Acid Sodium Salt, 25 G
SDP
Catalog No. 5000661088
Encompass_Preferred
Nalidixic Acid Sodium Salt, available in a 25g laboratory-grade powder form, is an effective antibacterial agent used to inhibit bacterial DNA replication. It is commonly used in microbiology laboratories for the detection and identification of Gram-negative bacteria, particularly in the case of urinary tract infections caused by Escherichia coli and other enteric pathogens. Nalidixic acid sodium salt is also used for antimicrobial susceptibility testing and as a supplement in selective media formulations. The 25g powder format allows for easy handling and accurate preparation of antibiotic solutions for research and diagnostic purposes.
Catalog No. 50-006-61088 Supplier Alkali Scientific Supplier No. ALI002CP
Nalidixic Acid Sodium Salt, available in a 25g laboratory-grade powder form, is an effective antibacterial agent used to inhibit bacterial DNA replication. It is commonly used in microbiology laboratories for the detection and identification of Gram-negative bacteria, particularly in the case of urinary tract infections caused by Escherichia coli and other enteric pathogens. Nalidixic acid sodium salt is also used for antimicrobial susceptibility testing and as a supplement in selective media formulations. The 25g powder format allows for easy handling and accurate preparation of antibiotic solutions for research and diagnostic purposes.