Learn More
Alkali Scientific Nalidixic Acid Sodium Salt, 5 G
SDP
Catalog No. 5000661089
Encompass_Preferred
Nalidixic Acid Sodium Salt, in a 5g laboratory-grade powder form, is used for bacterial inhibition by interfering with DNA synthesis in Gram-negative bacteria. It is commonly utilized in clinical microbiology for testing bacterial susceptibility and isolating pathogens, particularly those causing urinary tract infections. Nalidixic acid sodium salt is a critical tool in the identification of antibiotic-resistant strains and is frequently added to culture media for selective bacterial growth. The 5g powder format allows for accurate dosing and preparation of solutions, making it suitable for research applications and routine microbiological testing in laboratory settings.
Catalog No. 50-006-61089 Supplier Alkali Scientific Supplier No. ALI003CP
Nalidixic Acid Sodium Salt, in a 5g laboratory-grade powder form, is used for bacterial inhibition by interfering with DNA synthesis in Gram-negative bacteria. It is commonly utilized in clinical microbiology for testing bacterial susceptibility and isolating pathogens, particularly those causing urinary tract infections. Nalidixic acid sodium salt is a critical tool in the identification of antibiotic-resistant strains and is frequently added to culture media for selective bacterial growth. The 5g powder format allows for accurate dosing and preparation of solutions, making it suitable for research applications and routine microbiological testing in laboratory settings.