Learn More
Research Products International Corp Nalidixic Acid Sodium Salt, 25 Grams
SDP
Catalog No. 50997933
Encompass_Preferred
Nalidixic Acid Sodium Salt is a compound of notable importance in microbiology and pharmaceutical research. This sodium salt formulation of nalidixic acid is a valuable tool for investigating bacterial resistance mechanisms and antibiotic development. 
In microbiology, Nalidixic Acid Sodium Salt is widely employed to study bacterial susceptibility and resistance patterns. It serves as a selective agent in microbiological culture media, inhibiting the growth of susceptible bacteria. Researchers use it to assess the effectiveness of antibiotics and to understand the molecular mechanisms behind bacterial resistance, contributing to our understanding of microbial biology and antibiotic resistance. 
Catalog No. 50-997-933 Supplier Research Products International Corp Supplier No. N2310025.0
Nalidixic Acid Sodium Salt is a compound of notable importance in microbiology and pharmaceutical research. This sodium salt formulation of nalidixic acid is a valuable tool for investigating bacterial resistance mechanisms and antibiotic development. 
In microbiology, Nalidixic Acid Sodium Salt is widely employed to study bacterial susceptibility and resistance patterns. It serves as a selective agent in microbiological culture media, inhibiting the growth of susceptible bacteria. Researchers use it to assess the effectiveness of antibiotics and to understand the molecular mechanisms behind bacterial resistance, contributing to our understanding of microbial biology and antibiotic resistance. 