Learn More
Medchemexpress LLC Nalidixic acid (Standard) | 389-08-2 | 100.0% | 100 MG
SDP
Catalog No. 5000406411
Encompass_Preferred
Nalidixic acid (Standard) is the analytical standard of Nalidixic acid, intended for research and analytical applications. It is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria, acting in a bacteriostatic manner at lower concentrations and bactericidal at higher concentrations. It functions by inhibiting a subunit of DNA gyrase and topoisomerase IV, reversibly blocking DNA replication in susceptible bacteria.
  • Analytical standard
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits DNA gyrase and topoisomerase IV
  • Blocks DNA replication
  • Used as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-06411 Supplier Medchemexpress LLC Supplier No. HYB0398R100MG
Add to Cart
Add to Cart
Nalidixic acid (Standard) is the analytical standard of Nalidixic acid, intended for research and analytical applications. It is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria, acting in a bacteriostatic manner at lower concentrations and bactericidal at higher concentrations. It functions by inhibiting a subunit of DNA gyrase and topoisomerase IV, reversibly blocking DNA replication in susceptible bacteria.
  • Analytical standard
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits DNA gyrase and topoisomerase IV
  • Blocks DNA replication
  • Used as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS