Learn More
Medchemexpress LLC Nalidixic acid (Standard) | 389-08-2 | 99.98% | 50 MG
SDP
Catalog No. 5000407324
Encompass_Preferred
Nalidixic acid (Standard) is the analytical standard of Nalidixic acid, intended for research and analytical applications. It is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria, acting in a bacteriostatic manner at lower concentrations and bactericidal at higher concentrations. It inhibits a subunit of DNA gyrase and topoisomerase IV, reversibly blocking DNA replication in susceptible bacteria.
  • Analytical standard for research and analytical applications.
  • Quinolone antibiotic effective against gram-positive and gram-negative bacteria.
  • Acts as both bacteriostatic (lower concentrations) and bactericidal (higher concentrations).
  • Inhibits a subunit of DNA gyrase and topoisomerase IV.
  • Reversibly blocks DNA replication in susceptible bacteria.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-07324 Supplier Medchemexpress LLC Supplier No. HYB0398R50MG
Add to Cart
Add to Cart
Nalidixic acid (Standard) is the analytical standard of Nalidixic acid, intended for research and analytical applications. It is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria, acting in a bacteriostatic manner at lower concentrations and bactericidal at higher concentrations. It inhibits a subunit of DNA gyrase and topoisomerase IV, reversibly blocking DNA replication in susceptible bacteria.
  • Analytical standard for research and analytical applications.
  • Quinolone antibiotic effective against gram-positive and gram-negative bacteria.
  • Acts as both bacteriostatic (lower concentrations) and bactericidal (higher concentrations).
  • Inhibits a subunit of DNA gyrase and topoisomerase IV.
  • Reversibly blocks DNA replication in susceptible bacteria.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.