Learn More
ATCC NALM-1; Lymphoblast; Human (Homo sapiens)
Catalog No. 502383281
Encompass
NALM-1 is a lymphoblast cell line that was isolated from the peripheral blood of a three-year-old, female patient with leukemia. This cell line was deposited by J Minowada and can be used in immunology and immune system disorder research.
Catalog No. 50-238-3281 Supplier ATCC Supplier No. CRL1567
NALM-1 is a lymphoblast cell line that was isolated from the peripheral blood of a three-year-old, female patient with leukemia. This cell line was deposited by J Minowada and can be used in immunology and immune system disorder research.