Learn More
ATCC NALM6, clone G5; Lymphoma; Human (Homo sapiens)
Catalog No. 502384114
Encompass
NALM6, clone G5 is a lymphocyte-like that was isolated from the peripheral blood of a 19-year-old, male patient with acute lymphoblastic leukemia. This cell line was deposited by RS (Treichel) Salter (Oberlin College) in 2010.
Catalog No. 50-238-4114 Supplier ATCC Supplier No. CRL3273
NALM6, clone G5 is a lymphocyte-like that was isolated from the peripheral blood of a 19-year-old, male patient with acute lymphoblastic leukemia. This cell line was deposited by RS (Treichel) Salter (Oberlin College) in 2010.