Learn More
Cayman Chemical Nalmefene (hydrochloride), Purity: ≥98%, CAS: 58895-64-0, 50 mg
SDP
Catalog No. 502941543
Encompass_Preferred
A μ- and κ-opioid receptor antagonist (Kis = 0.91 and 1.03 nM, respectively, in CHO membranes expressing the human receptors); selective for μ- and κ-opioid receptors over δ-opioid receptors (Ki = 13.26 nM in CHO membranes expressing the human receptors); decreases self-administration of alcohol in non-alcohol- and alcohol-dependent rats; reduces food and water intake and body weight in obese and lean rats at 25 mg/kg per day; inhibits morphine-induced scratching behavior and increases in the latency to tail withdrawal in a warm water tail withdrawal assay in rhesus monkeys at 32 µg/kg
Catalog No. 50-294-1543 Supplier Cayman Chemical Supplier No. 2826650 MG
A μ- and κ-opioid receptor antagonist (Kis = 0.91 and 1.03 nM, respectively, in CHO membranes expressing the human receptors); selective for μ- and κ-opioid receptors over δ-opioid receptors (Ki = 13.26 nM in CHO membranes expressing the human receptors); decreases self-administration of alcohol in non-alcohol- and alcohol-dependent rats; reduces food and water intake and body weight in obese and lean rats at 25 mg/kg per day; inhibits morphine-induced scratching behavior and increases in the latency to tail withdrawal in a warm water tail withdrawal assay in rhesus monkeys at 32 µg/kg