Learn More
Medchemexpress LLC Naloxegol oxalate | 1354744-91-4 | 99.9% | C36H55NO15 | 5 MG
SDP
Catalog No. 5000432638
Encompass_Preferred
Naloxegol oxalate is an orally active, peripherally acting μ-opioid receptor antagonist (target Ki 7.42 nmol/L). It inhibits opioid binding to gastrointestinal μ-opioid receptors, alleviating opioid-induced gastrointestinal hypomotility, delayed transit, hypertonicity, and increased fluid reabsorption. It is applicable for research into opioid-induced constipation.
  • Orally active
  • Peripherally acting μ-opioid receptor antagonist
  • Inhibits opioid binding to gastrointestinal μ-opioid receptors
  • Alleviates opioid-induced gastrointestinal hypomotility and related issues
  • Applicable to research on opioid-induced constipation
  • High affinity binding to μ and κ-opioid receptors in vitro
  • Low affinity to δ-opioid receptors in vitro
  • No significant activity at non-opioid targets in vitro
  • Available in solid and solution forms
Catalog No. 50-004-32638 Supplier Medchemexpress LLC Supplier No. HYA0118A5MG
Naloxegol oxalate is an orally active, peripherally acting μ-opioid receptor antagonist (target Ki 7.42 nmol/L). It inhibits opioid binding to gastrointestinal μ-opioid receptors, alleviating opioid-induced gastrointestinal hypomotility, delayed transit, hypertonicity, and increased fluid reabsorption. It is applicable for research into opioid-induced constipation.
  • Orally active
  • Peripherally acting μ-opioid receptor antagonist
  • Inhibits opioid binding to gastrointestinal μ-opioid receptors
  • Alleviates opioid-induced gastrointestinal hypomotility and related issues
  • Applicable to research on opioid-induced constipation
  • High affinity binding to μ and κ-opioid receptors in vitro
  • Low affinity to δ-opioid receptors in vitro
  • No significant activity at non-opioid targets in vitro
  • Available in solid and solution forms