Learn More
Medchemexpress LLC Naloxegol oxalate | 1354744-91-4 | 99.9% | C36H55NO15 | 50 MG
SDP
Catalog No. 5000432654
Encompass_Preferred
Naloxegol oxalate is an orally active, peripherally acting μ-opioid receptor antagonist. It works by inhibiting the binding of opioids to μ-opioid receptors in the gastrointestinal tract, thereby alleviating opioid-induced conditions like hypomotility, delayed transit, hypertonicity, and increased fluid reabsorption. This compound is primarily used in research related to opioid-induced constipation.
  • Orally active, peripherally acting μ-opioid receptor antagonist
  • Inhibits opioid binding in the gastrointestinal tract
  • Alleviates opioid-induced gastrointestinal hypomotility
  • Used in research for opioid-induced constipation
  • Binds with high affinity to μ-opioid and κ-opioid receptors
  • Shows low affinity to δ-opioid receptors with no significant activity at non-opioid targets
Catalog No. 50-004-32654 Supplier Medchemexpress LLC Supplier No. HYA0118A50MG
Naloxegol oxalate is an orally active, peripherally acting μ-opioid receptor antagonist. It works by inhibiting the binding of opioids to μ-opioid receptors in the gastrointestinal tract, thereby alleviating opioid-induced conditions like hypomotility, delayed transit, hypertonicity, and increased fluid reabsorption. This compound is primarily used in research related to opioid-induced constipation.
  • Orally active, peripherally acting μ-opioid receptor antagonist
  • Inhibits opioid binding in the gastrointestinal tract
  • Alleviates opioid-induced gastrointestinal hypomotility
  • Used in research for opioid-induced constipation
  • Binds with high affinity to μ-opioid and κ-opioid receptors
  • Shows low affinity to δ-opioid receptors with no significant activity at non-opioid targets