Learn More
Medchemexpress LLC Naloxegol oxalate | 1354744-91-4 | 99.91% | C36H55NO15 | 100 MG
SDP
Catalog No. 5000432697
Encompass_Preferred
Naloxegol oxalate is an orally active, peripherally acting μ-opioid receptor antagonist. It inhibits the binding of opioids to μ-opioid receptors in the gastrointestinal tract, alleviating opioid-induced gastrointestinal hypomotility, delayed transit, hypertonicity, and increased fluid reabsorption. It is used in research related to opioid-induced constipation.
  • Binds with high affinity to μ-opioid and κ-opioid receptors
  • Exhibits low affinity to δ-opioid receptors in vitro
  • Shows no significant activity at non-opioid targets
  • Alleviates opioid-induced slowed gastrointestinal motility and transit
Catalog No. 50-004-32697 Supplier Medchemexpress LLC Supplier No. HYA0118A100MG
Naloxegol oxalate is an orally active, peripherally acting μ-opioid receptor antagonist. It inhibits the binding of opioids to μ-opioid receptors in the gastrointestinal tract, alleviating opioid-induced gastrointestinal hypomotility, delayed transit, hypertonicity, and increased fluid reabsorption. It is used in research related to opioid-induced constipation.
  • Binds with high affinity to μ-opioid and κ-opioid receptors
  • Exhibits low affinity to δ-opioid receptors in vitro
  • Shows no significant activity at non-opioid targets
  • Alleviates opioid-induced slowed gastrointestinal motility and transit