Learn More
Medchemexpress LLC Naloxegol oxalate | 1354744-91-4 | MFCD29472292 | 99.9% | 741.82 | C36H55NO15 | 10 MG
SDP
Catalog No. 5000298776
Encompass_Preferred
Naloxegol oxalate is the oxalate salt of naloxegol, a peripherally selective μ-opioid receptor antagonist used in research to study and mitigate opioid-induced constipation. The product is supplied as a high-purity solid and as a DMSO solution for in vitro and preclinical applications.
  • Peripherally selective μ-opioid receptor antagonist.
  • High purity (99.9%), suitable for analytical and biological assays.
  • Available as 10 mg solid packs and as 10 mM solution in DMSO.
  • Commonly used to study opioid-induced gastrointestinal effects and receptor pharmacology.
  • Chemical identifiers: CAS 1354744-91-4; molecular weight 741.82.
Catalog No. 50-002-98776 Supplier Medchemexpress LLC Supplier No. HYA0118A10MG
Naloxegol oxalate is the oxalate salt of naloxegol, a peripherally selective μ-opioid receptor antagonist used in research to study and mitigate opioid-induced constipation. The product is supplied as a high-purity solid and as a DMSO solution for in vitro and preclinical applications.
  • Peripherally selective μ-opioid receptor antagonist.
  • High purity (99.9%), suitable for analytical and biological assays.
  • Available as 10 mg solid packs and as 10 mM solution in DMSO.
  • Commonly used to study opioid-induced gastrointestinal effects and receptor pharmacology.
  • Chemical identifiers: CAS 1354744-91-4; molecular weight 741.82.