Learn More
TARGETMOL CHEMICALS INC Naloxegol oxalate 50MG
SDP
Catalog No. 502879497
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Naloxegol oxalate (NKTR-118) is a peripherally-selective opioid antagonist for the treatment of opioid-induced constipation. Purity 99.82%
Catalog No. 50-287-9497 Supplier TARGETMOL CHEMICALS INC Supplier No. T335550MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Naloxegol oxalate (NKTR-118) is a peripherally-selective opioid antagonist for the treatment of opioid-induced constipation. Purity 99.82%