Learn More
Medchemexpress LLC Naloxone | 465-65-6 | 99.7% | 327.37 | 200 MG
SDP
Catalog No. 5000384307
Encompass_Preferred
Naloxone is an opioid receptor antagonist that alleviates opioid-overdose-induced respiratory depression. It may cause pulmonary edema and cardiac arrhythmias and is intended for research use only, not for sale to patients.
  • Antagonist of opioid receptors
  • Alleviates respiratory depression
  • Soluble in DMSO (100 mg/mL)
  • Stable at 4°C, protected from light, under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Suitable for in vitro and in vivo research applications
  • Demonstrates activity in CHO and HEK293 cells
  • Effective in animal models, including rats and cats
Catalog No. 50-003-84307 Supplier Medchemexpress LLC Supplier No. HY17417A200MG
Naloxone is an opioid receptor antagonist that alleviates opioid-overdose-induced respiratory depression. It may cause pulmonary edema and cardiac arrhythmias and is intended for research use only, not for sale to patients.
  • Antagonist of opioid receptors
  • Alleviates respiratory depression
  • Soluble in DMSO (100 mg/mL)
  • Stable at 4°C, protected from light, under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Suitable for in vitro and in vivo research applications
  • Demonstrates activity in CHO and HEK293 cells
  • Effective in animal models, including rats and cats