Learn More
U.S. Pharmacopeia Naloxone, 465-65-6, MFCD00242634, 200 mg
Catalog No. NC1364637
Encompass
Empirical Formula (Hill Notation): C19H21NO4, Molecular Weight: 327.37, Synonym: (5α)-4,5-Epoxy-3,14-dihydroxy-17-(2-propen-1-yl)morphinan-6-one.
Catalog No. NC1364637 Supplier U.S. Pharmacopeia Supplier No. 1453005
Empirical Formula (Hill Notation): C19H21NO4, Molecular Weight: 327.37, Synonym: (5α)-4,5-Epoxy-3,14-dihydroxy-17-(2-propen-1-yl)morphinan-6-one.