Learn More
Medchemexpress LLC Naloxone hydrochloride | 357-08-4 | MFCD00069322 | 99.7% | 363.84 g/mol | C19H22ClNO4 | 10 MG
SDP
Catalog No. 5000298161
Encompass_Preferred
Naloxone hydrochloride is a research-grade reference standard of naloxone, a competitive opioid receptor antagonist used to reverse opioid effects and to support pharmacology and analytical method development. Supplied as a 10 mg reference material with accompanying certificate of analysis and safety data sheet, it is intended for research and analytical applications.
  • High purity suitable for analytical and reference applications (99.7%).
  • Provided with certificate of analysis and safety data sheet.
  • Suitable for opioid receptor pharmacology and receptor-binding studies.
  • Molecular weight 363.84 g/mol; formula C19H22ClNO4.
  • Packaged as a 10 mg reference standard for method development and validation.
Catalog No. 50-002-98161 Supplier Medchemexpress LLC Supplier No. HY17417R10MG
Naloxone hydrochloride is a research-grade reference standard of naloxone, a competitive opioid receptor antagonist used to reverse opioid effects and to support pharmacology and analytical method development. Supplied as a 10 mg reference material with accompanying certificate of analysis and safety data sheet, it is intended for research and analytical applications.
  • High purity suitable for analytical and reference applications (99.7%).
  • Provided with certificate of analysis and safety data sheet.
  • Suitable for opioid receptor pharmacology and receptor-binding studies.
  • Molecular weight 363.84 g/mol; formula C19H22ClNO4.
  • Packaged as a 10 mg reference standard for method development and validation.