Learn More
Enzo Life Sciences Naloxone . hydrochloride (50mg). CAS: 357-08-4
SDP
Catalog No. 502009468
Encompass_Preferred
Opioid antagonist. Alternative name: (5alpha)-17-allyl-4,5-epoxy-3,14-dihydroxymorphinan-6-one hydrochloride. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-9468 Supplier Enzo Life Sciences Supplier No. ALX550055M050
Opioid antagonist. Alternative name: (5alpha)-17-allyl-4,5-epoxy-3,14-dihydroxymorphinan-6-one hydrochloride. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water or 100% ethanol. Long Term Storage: +4°C.