Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naloxone Hydrochloride, Dihydrate, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604724 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
300 g
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-724 250 mg Amber Glass Bottle
18-604-725 1 g Amber Glass Bottle
18-604-726 5 g Amber Glass Bottle
18-604-727 25 g Amber Glass Bottle
18-604-728 300 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-724 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1231250MG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

N1231, 51481-60-8, C19H21NO4·HCl·2H2O

Spectrum™ Chemical Naloxone Hydrochloride, Dihydrate, USP is an opioid antagonist used for the complete or partial reversal of opioid overdose. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51481-60-8

Specifications

CAS Max % 1
Loss on Drying 0.11
Packaging Amber Glass Bottle
Quantity 250 mg
Specific Rotation -170° to -181°
Molecular Weight (g/mol) 399.87
Percent Purity 98 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Naloxone Hydrochloride Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.