Learn More
Medchemexpress LLC Naloxone (hydrochloride) (Standard) | 357-08-4 | 100.0% | 363.84 | 100 MG
SDP
Catalog No. 5000386103
Encompass_Preferred
Naloxone hydrochloride is an Opioid receptor antagonist that alleviates opioid-overdose-induced respiratory depression and may cause pulmonary edema and cardiac arrhythmias. It has a molecular weight of 363.84 and a chemical formula of C19H22ClNO4.
  • Appears as a solid, white to off-white substance
  • Soluble in water at 62.5 mg/mL (requires ultrasonic assistance)
  • Soluble in DMSO at concentrations of 30 mg/mL or greater
  • Typically shipped at room temperature within the continental US
  • Store solid product at 4°C in sealed containers, away from moisture
  • Store solutions in solvent at -80°C for up to 6 months or -20°C for 1 month
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
Catalog No. 50-003-86103 Supplier Medchemexpress LLC Supplier No. HY17417R100MG
Naloxone hydrochloride is an Opioid receptor antagonist that alleviates opioid-overdose-induced respiratory depression and may cause pulmonary edema and cardiac arrhythmias. It has a molecular weight of 363.84 and a chemical formula of C19H22ClNO4.
  • Appears as a solid, white to off-white substance
  • Soluble in water at 62.5 mg/mL (requires ultrasonic assistance)
  • Soluble in DMSO at concentrations of 30 mg/mL or greater
  • Typically shipped at room temperature within the continental US
  • Store solid product at 4°C in sealed containers, away from moisture
  • Store solutions in solvent at -80°C for up to 6 months or -20°C for 1 month
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation