Learn More
Medchemexpress LLC Naloxone hydrochloride (standard) | 357-08-4 | 99.7% | 363.84 | 50 MG
SDP
Catalog No. 5000384024
Encompass_Preferred
Naloxone hydrochloride (Standard) is the analytical standard of Naloxone hydrochloride, intended for research and analytical applications. It functions as an antagonist of the Opioid receptor and is known to alleviate opioid-overdose-induced respiratory depression. This compound is a reference standard supplied for assay and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used for research and analytical applications.
  • Functions as an Opioid receptor antagonist.
  • Alleviates opioid-overdose-induced respiratory depression.
  • Suitable for HPLC, GC, and MS experiments.
  • Reference standard grade for assay.
Catalog No. 50-003-84024 Supplier Medchemexpress LLC Supplier No. HY17417R50MG
Naloxone hydrochloride (Standard) is the analytical standard of Naloxone hydrochloride, intended for research and analytical applications. It functions as an antagonist of the Opioid receptor and is known to alleviate opioid-overdose-induced respiratory depression. This compound is a reference standard supplied for assay and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used for research and analytical applications.
  • Functions as an Opioid receptor antagonist.
  • Alleviates opioid-overdose-induced respiratory depression.
  • Suitable for HPLC, GC, and MS experiments.
  • Reference standard grade for assay.