Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naloxone Hydrocloride, MP Biomedicals™

Catalog No. MP219024580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219024580 100 mg
MP219024525 25 mg
MP219024583 250 mg
MP219024591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219024580 Supplier MP Biomedicals, Inc Supplier No. 0219024580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naloxone Hydrocloride Off-white Crystalline Powder

  • The role of naloxone (1μM) and other opioids on apoptosis in several human cancer cell lines has been probed. Naloxone (10-100μM) has been used to study proliferation and neurogenesis in cultured rat adult hippocampal progenitors.
  • Naloxone has also been utilized to modulate the NMDA receptor activation capability of theμ-opioid receptor agonist DAMGO. It displaces morphine and other opiate agonists from the opiate receptor.
  • It is more potent than Nalorphine and is also an effective antagonist for mixed agonist-antagonist drugs such as pentazorcine. Naloxone has no respiratory depressant activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357-08-4
Molecular Formula C19H22ClNO4
Molecular Weight (g/mol) 363.84
InChI Key RGPDIGOSVORSAK-STHHAXOLSA-N
Synonym Narcan, EN-1530, (5α)-4,5-Epoxy-3,14-dihydroxy-17-(2-propenyl)morphinan-6-one hydrochloride, 17-allyl-4,5α- epoxy-3,14-dihydroxymorphinan-6-one hydrochloride
PubChem CID 5464092

Specifications

Melting Point 129.0°C to 129.2°C (literature)
Color Off-white
Quantity 100 mg
Beilstein 6260807
Solubility Information Soluble in water (50mg/mL - clear, colorless to faint yellow solution), ethanol (3.3mg/mL), or methanol (50mg/mL - clear, colorless solution).
Physical Form Crystalline Powder
RTECSNumber QD2275000
storageNote1 Store at +4°C, Store Desiccated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.