Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Naloxone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 51481-60-8 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C19H21 NO4 · HCl · 2H2 O |
| MDL Number | MFCD00150901 |
| Synonym | Naloxone hydrochloride dihydrate |
| Type | Certified Reference Material |
| Quantity | 300 mg |
| Formula Weight | 399.87 |
| Grade | Certified Reference Material |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?