Learn More
Cayman Chemical Naloxone (hydrochloride), Purity: ≥95%, CAS: 357-08-4, 50 mg
SDP
Catalog No. 502972631
Encompass_Preferred
An analytical reference standard categorized as an opioid antagonist; formulations containing it have been used as antidotes for opioid overdose and the prevention of overdose; formulations containing it have also been used in the treatment of opiate addiction and in pain management; intended for research and forensic applications
Catalog No. 50-297-2631 Supplier Cayman Chemical Supplier No. 1559450 MG
An analytical reference standard categorized as an opioid antagonist; formulations containing it have been used as antidotes for opioid overdose and the prevention of overdose; formulations containing it have also been used in the treatment of opiate addiction and in pain management; intended for research and forensic applications