Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Nalpha-4-Tosyl-L-arginine methyl ester hydrochloride, 98+%

Catalog No. AC139200500
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139200500 50 g Glass bottle
AC139200050 5 g Glass bottle
AC139200100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139200500 Supplier Thermo Scientific Chemicals Supplier No. 139200500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1784-03-8 | C14H22N4O4S·HCl | 378.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1784-03-8
Molecular Formula C14H22N4O4S·HCl
Molecular Weight (g/mol) 378.88
MDL Number MFCD00012578
InChI Key JIQFFACVQXXHMY-YDALLXLXSA-N
Synonym tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
PubChem CID 2723792
IUPAC Name methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 146°C to 149°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NHSO2C6H4CH3)CO2CH3
Quantity 50 g
Specific Rotation -14
Formula Weight 378.88
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nα-4-Tosyl-L-arginine methyl ester hydrochloride
Show More Show Less
EINECSNumber 217-235-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.