Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide 98.0+%, TCI America™
SDP

Catalog No. A5524100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5524100MG 100 mg
A55241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5524100MG Supplier TCI America Supplier No. A5524100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 178065-30-0
Molecular Formula C12H15FN4O5
Molecular Weight (g/mol) 314.27
MDL Number MFCD03844762
InChI Key WCOZOJGXDVGGIK-UHFFFAOYNA-N
Synonym Nalpha-(2,4-Dinitro-5-fluorophenyl)-D-leucinamide
PubChem CID 15817227
IUPAC Name 2-[(5-fluoro-2,4-dinitrophenyl)amino]-4-methylpentanamide
SMILES CC(C)CC(NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

Specifications

Melting Point 170°C
Color Yellow
UN Number 1325
Quantity 100 mg
Formula Weight 314.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide [HPLC Labeling Reagent for e.e. Determination]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.