Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester 98.0+%, TCI America™
SDP

Catalog No. F05171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05171G 1 g
F05175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05171G Supplier TCI America Supplier No. F05171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120791-76-6
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.47
MDL Number MFCD08276378
InChI Key LMRQPSBMLDCJGN-UHFFFAOYNA-N
Synonym Nalpha-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu
PubChem CID 11773868
IUPAC Name tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoate
SMILES CC(O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C

Specifications

Melting Point 100°C
Color White
Quantity 1 g
Formula Weight 397.47
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.