Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-tyrosine 98.0+%, TCI America™
SDP

Catalog No. F07721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07721G 1 g
F07725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07721G Supplier TCI America Supplier No. F07721G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 118488-18-9
Molecular Formula C28H29NO5
Molecular Weight (g/mol) 459.54
MDL Number MFCD00065684
InChI Key JAUKCFULLJFBFN-KSYWNVGFNA-N
Synonym fmoc-d-tyr tbu-oh, fmoc-o-tert-butyl-d-tyrosine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, nalpha-9h-fluoren-9-ylmethoxy carbonyl-o-tert-butyl-d-tyrosine, 2r-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-tyrosine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, ambotzfaa1341, pubchem10054, fmoc-d-tyr but-oh, fmoc-o-tert.butyl-d-tyrosine
PubChem CID 12968135
IUPAC Name (2R)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Specifications

Melting Point 152°C
Color White
Quantity 1 g
Formula Weight 459.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-tyrosine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.