Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-threonine 98.0+%, TCI America™
SDP

Catalog No. F05055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05055G 5 g
F05051G 1 g
F050525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F05055G Supplier TCI America Supplier No. F05055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71989-35-0
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.47
MDL Number MFCD00077075
InChI Key LZOLWEQBVPVDPR-VLIAUNLRSA-N
Synonym fmoc-thr tbu-oh, fmoc-o-tert-butyl-l-threonine, fmoc-thr t-bu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, fmoc-thr but, 2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine, fmoc-thr but-oh, fmoc-o-tert.butyl-l-threonine, fmoc-thr otbu-oh
PubChem CID 6364643
IUPAC Name (2S,3R)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 5 g
Formula Weight 397.47
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.