Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Acetyl-L-glutamine 99.0+%, TCI America™
SDP

Catalog No. A078425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A078425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A078425G Supplier TCI America Supplier No. A078425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent with a minimum 99.0% titration purity.

Chemical Identifiers

CAS 2490-97-3
Molecular Formula C7H12N2O4
Molecular Weight (g/mol) 188.183
MDL Number MFCD00038159
InChI Key KSMRODHGGIIXDV-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
PubChem CID 182230
ChEBI CHEBI:21553
IUPAC Name (2S)-2-acetamido-5-amino-5-oxopentanoic acid
SMILES CC(=O)NC(CCC(=O)N)C(=O)O

Specifications

Melting Point 208°C
Color White
Quantity 25 g
Formula Weight 188.18
Percent Purity ≥99.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Acetyl-L-glutamine
EINECSNumber (9)-1582
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.