Learn More
Sigma Aldrich Fine Chemicals Biosciences Nalpha Benzoyl L arginine1G
Catalog No. 502914428
Encompass
NBenzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.
Catalog No. 50-291-4428 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B31331G
NBenzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.