Learn More
Sigma Aldrich Fine Chemicals Biosciences Nalpha Benzoyl L arginine250MG
Catalog No. 502903972
Encompass
NBenzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.
Catalog No. 50-290-3972 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B3133250MG
NBenzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.