Learn More
Sigma Aldrich Fine Chemicals Biosciences Nalpha-Benzoyl-L-arginine25MG
Catalog No. 502899029
Encompass
NBenzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.
Catalog No. 50-289-9029 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B313325MG
NBenzoyl-L-arginine 4-nitroanilide hydrochloride is a substrate for trypsin.